missing translation for 'onlineSavingsMsg'
Learn More

Phenacetin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012228
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012228 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012228 Supplier Merck Emd Millipore Supplier No. PHR12491G

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 62-44-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Linear Formula CH3 CONHC6H4 OC2H5
MDL Number MFCD00009094
Melting Point 133°C to 136°C
Synonym 1-Acetyl-p-phenetidin; 4′-Ethoxyacetanilide; N-(4-Ethoxyphenyl)acetamide; p-Acetophenetidide; Acetophenetidin
Type Certified Reference Material
Quantity 1 g
Formula Weight 179.22
Grade Certified Reference Material