missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phe-Arg-β-naphthylamide dihydrochloride, >99%, For TLC analysis, 85.4%, MP Biomedicals™
$270.96 - $270.96
Specifications
| Purity | >99% |
|---|---|
| Assay | 0.854 |
| CAS | 100929-99-5 |
| Color | White |
| Content And Storage | Store at 0°C. |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15612825
|
MP Biomedicals Inc
0215612825 |
25mg |
Each for $270.96
|
|
|||||
Specifications
| >99% | |
| 100929-99-5 | |
| Store at 0°C. | |
| 519.471 |
| 0.854 | |
| White | |
| Powder | |
| Phe-Arg-β-naphthylamide dihydrochloride |