accessibility menu, dialog, popup

UserName

CAS RN 100929-99-5

IUPAC Name: 2-(2-amino-3-phenylpropanamido)-5-[(diaminomethylidene)amino]-N-(naphthalen-2-yl)pentanamide dihydrochloride
Synonyms: 2-(2-amino-3-phenylpropanamido)-5-[(diaminomethylidene)amino]-N-(naphthalen-2-yl)pentanamide dihydrochloride
Molecular Weight (g/mol): 519.47
Molecular Formula: C25H32Cl2N6O2
InChi Key: MRUMOHLDHMZGMS-UHFFFAOYNA-N
SMILES: Cl.Cl.NC(CC1=CC=CC=C1)C(=O)NC(CCCN=C(N)N)C(=O)NC1=CC=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results