missing translation for 'onlineSavingsMsg'
Learn More

PHC3 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501560721
Encompass
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501560721 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501560721 Supplier Bethyl Laboratories Inc Supplier No. A301569AT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

Component of a Polycomb group (PcG) multiprotein PRC1-like complex, a complex class required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A ′Lys-119′, rendering chromatin heritably changed in its expressibility.

Specifications

Antigen PHC3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PHC3
Gene Accession No. Q8NDX5
Gene Alias EDR3, HPH3, EDR3, PH3, Polyhomeotic-like protein 3, Early development regulatory protein 3, hPH3
Gene Symbols PHC3
Host Species Rabbit
Immunogen Between 700 and 750
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80012
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less