Learn More
Pharmaceutical Analytics QuantStudio™ 5 Real-Time PCR System, 96-well, 0.1 mL, laptop
Description
The Applied Biosystems™ QuantStudio™ 5 Real-Time PCR System paired with Pharmaceutical Analytics AccuSEQ v3.1 is designed for users who need superior performance and security options in a real-time PCR system that is affordable and easy to use. The QuantStudio 5 pharmaceutical analytics system is used together with specialized sample preparation and quantitation kits for host cell protein, host cell DNA, and Protein A impurity testing of biological products produced in cell culture. The QuantStudio 5 system also offers the built-in software features of electronic record security and prevention of unauthorized instrument access to assist with 21 CFR Part 11 compliance.
Features of the QuantStudio 5 system with 96-well block include:
- Validated for use with AccuSEQ software version 3.1
- Integrated workflow for impurity and contaminant testing during biopharmaceutical manufacturing using the SEQ family of real-time PCR assays, including MycoSEQ, resDNASEQ, and ProteinSEQ assays
- User account management for greater control of experiment data
- Built-in software features to support compliance with 21 CFR Part 11 guidelines
- Simplified instrument software with interactive touchscreen for greater ease of use
- 96-well 0.1-mL format
Easy to use
The QuantStudio 5 Real-Time PCR System is compact in form, factory calibrated and is delivered in user-friendly packaging to enable fast and simple installation for immediate use. The interactive touchscreen provides a modern usability experience and the AccuSEQ Software v3.1 protocol templates allow users of any experience level to quickly set up and start a run.
Validated with AccuSEQ Software v3.1
- Full workflow SAE coverage with compliance services to help customer with regulatory requirements
- Available compliance services, including CSV, IQ/OQ/PV
- Validated on the latest 2019 WIN10 IOT operating system
- Includes latest security patches, non-forced auto updates
Total control
With built-in software features to support compliance with 21 CFR Part 11 guidelines, user account management, locked workflow and Pause features, the QuantStudio 5 system offers users greater control of experiment data.
Multiplex performance results that you can trust
Improved well-to-well and instrument-to-instrument data accuracy is achieved with OptiFlex technology. In the 96-well system this technology features 6 decoupled excitation and emission filter channels, for a total of 21 filter combinations, with white LED.
Sensitive data analysis
Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.
High application versatility
Simple and optimized protocols, reliable assays and reagents, and intuitive software are available for a broad range of applications, including gene expression, genetic variation, gene regulation, and more.
Specifications
Specifications
| Block Format | Non-interchangeable |
| Format | 96-well Plate |
| Content And Storage | QuantStudio 5 Real Time PCR System, 0.1 mL, 96-well, Laptop |
| Display Type | Touchscreen |
| Shipping Condition | Room Temperature |
| Quantity | 1 system |
Frequently Asked Questions (FAQs)
We recommend trying different browsers (Google Chrome, Firefox, and Microsoft Edge) and/or removing cookies from your browser. If this does not resolve the issue, please contact Technical Support at techsupport@thermofisher.com.
If the admin PIN has not been changed, 1234 should work. If it does not work, you can also try 0000.
However, if the admin PIN has been changed and none of the above PINs work, follow the instructions in the QuantStudio 3 and 5 Real-Time PCR Systems manual, page 100.
For Research Use Only. Not for use in diagnostic procedures.