missing translation for 'onlineSavingsMsg'
Learn More

Pharmaceutical Analytics QuantStudio™ 3 Real-Time PCR System, 96-well, 0.2 mL

Catalog No. A31665
Encompass
Change view
Click to view available options
Quantity:
1 System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31665 1 System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A31665 Supplier Thermo Fisher Scientific Supplier No. A31665

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Block Format Non-interchangeable
Format 96-well Plate
Content And Storage • QuantStudio 3 Real Time PCR System
• 0.2 mL 96-well block
• (Computer not included)
Display Type Touchscreen
Shipping Condition Room Temperature
Quantity 1 System
Can I downgrade the firmware on my QuantStudio 5?

You can perform this action if you have access to an Admin account on the QuantStudio instrument and if it is compatible for the downgrade (newer instruments are not compatible with v1.3.3). If your instrument serial number is below the following, then it can be downgraded:
QuantStudio 5 96-well, 0.1 mL block - SN 272514140
QuantStudio 5 96-well, 0.2 mL block - SN 2725210507
QuantStudio 5 384 well block - SN 272533285
Please also check if the instrument is being used with any validated protocols that may require a particular firmware version to be compliant. If you need the firmware itself, you can download it here.
The instructions to downgrade the firmware are on p.4 of the Release Notes: QuantStudioTM 3 & 5 Touchscreen Instrument Operating Software Version 1.3.3. It's the same process as an upgrade.

What is the admin pin for QuantStudio 3 and 5 Real-Time PCR Systems?

If the admin PIN has not been changed, 1234 should work. If it does not work, you can also try 0000.
However, if the admin PIN has been changed and none of the above PINs work, follow the instructions in the QuantStudio 3 and 5 Real-Time PCR Systems manual, page 100.

For Research Use Only. Not for use in diagnostic procedures.