missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phalloidin Labeling Probes

Catalog No. R415
Encompass
Change view
Click to view available options
Color:
Blue
Far-red
Green
Near-infrared
None
Orange
Orange-red
Red
Red-orange
Violet
Yellow
Excitation Wavelength Range:
346/442 nm
405/450 nm
495/518 nm
496/516 nm
496/520 nm
531/554 nm
540/565 nm
555/565 nm
556/570 nm
558/569 nm
578/600 nm
581/609 nm
591/608 nm
632/647 nm
633/647 nm
650/668 nm
663/690 nm
679/702 nm
758/784 nm
None
Dye Type:
Alexa Fluor Plus 555
Alexa Fluor Plus 647
Alexa Fluor™ 350
Alexa Fluor™ 488
Alexa Fluor™ 532
Alexa Fluor™ 546
Alexa Fluor™ 555
Alexa Fluor™ 568
Alexa Fluor™ 594
Alexa Fluor™ 633
Alexa Fluor™ 635
Alexa Fluor™ 647
Alexa Fluor™ 660
Alexa Fluor™ 680
Alexa Fluor™ Plus 405
Alexa Fluor™ Plus 750
Biotin-XX
BODIPY™
FITC (Fluorescein)
Oregon Green™ 488
Phalloidin (unlabeled)
Texas Red™
TRITC
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Excitation Wavelength Range Dye Type
R415 Red-orange 540/565 nm TRITC
A22281 Blue 346/442 nm Alexa Fluor™ 350
A30104 Violet 405/450 nm Alexa Fluor™ Plus 405
A12379 Green 495/518 nm Alexa Fluor™ 488
O7466 Green 496/520 nm Oregon Green™ 488
F432 Green 496/516 nm FITC (Fluorescein)
A22282 Yellow 531/554 nm Alexa Fluor™ 532
A22283 Orange 556/570 nm Alexa Fluor™ 546
A34055 Orange 555/565 nm Alexa Fluor™ 555
A30106 Orange 555/565 nm Alexa Fluor Plus 555
B3475 Red 558/569 nm BODIPY™
A12380 Orange-red 578/600 nm Alexa Fluor™ 568
A12381 Red 581/609 nm Alexa Fluor™ 594
T7471 Red 591/608 nm Texas Red™
A22284 Far-red 632/647 nm Alexa Fluor™ 633
A34054 Far-red 633/647 nm Alexa Fluor™ 635
A22287 Far-red 650/668 nm Alexa Fluor™ 647
A30107 Far-red 650/668 nm Alexa Fluor Plus 647
A22285 Near-infrared 663/690 nm Alexa Fluor™ 660
A22286 Near-infrared 679/702 nm Alexa Fluor™ 680
A30105 Near-infrared 758/784 nm Alexa Fluor™ Plus 750
B7474 None None Biotin-XX
P3457 None None Phalloidin (unlabeled)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Catalog No. R415 Supplier Invitrogen™ Supplier No. R415
Only null left

Achieve precise and reliable F-actin staining with fluorescent and biotinylated phalloidins. Phalloidin conjugates are widely used in imaging applications to selectively label F-actin in a variety of sample types including fixed and permeabilized cells, tissue sections, and cell-free experiments.

Fluorescent and biotinylated phalloidins are water soluble and bind to filamentous actin (F-actin) with nanomolar affinity, making them convenient probes for labeling, identifying, and quantifying F-actin in cryopreserved tissue sections, fixed and permeabilized cells, and cell-free experiments. Phalloidin conjugates bind similarly to actin from various species, including plants and animals, enabling staining of the cytoskeleton in a wide range of samples.

A variety of phalloidin conjugates for filamentous (F-actin) staining are available, including fluorescent Alexa Fluor and Alexa Fluor Plus phalloidins, along with phalloidins conjugated to classic fluorescent dyes such as BODIPY, fluorescein, and rhodamine. Phalloidin staining is spectrally compatible with other fluorescent stains used in cellular analyses such as GFP/RFP, Qdot nanocrystals, and other Alexa Fluor conjugates and antibodies. Biotin‐XX Phalloidin can be used to visualize actin filaments via fluorescent streptavidin tags or standard enzyme-mediated avidin/streptavidin techniques such as in electron microscopy. Unlabeled phalloidin is available for use as a control in blocking F‐actin staining or in promoting polymerization.

Phalloidin conjugates bind to both large and small actin filaments with similar affinity in a 1:1 stoichiometry between phallotoxin and actin subunits. They do not bind G-actin monomers.

Alexa Fluor and Alexa Fluor Plus phalloidin conjugates for F-actin staining

Fluorescent Alexa Fluor dye conjugates of phalloidin are popular F-actin stains, offering color choices across the full spectral range. These phalloidin conjugates provide researchers with fluorescent probes that are superior in brightness and photostability compared to other spectrally similar conjugates.

Alexa Fluor Plus Phalloidin conjugates retain the same specificity for actin but offer 3-5 times greater sensitivity and brightness compared to the corresponding Alexa Fluor Phalloidin conjugate. This increased brightness is beneficial for challenging F-actin imaging, such as the super‐resolution microscopy methods SIM and STORM, and for reliable staining of actin stress fibers.

Features of phalloidin probes

  • High specificity—binds selectively to F-actin, which allows for precise labeling of actin filaments in fixed cells and cryopreserved tissues
  • Strong affinity—nanomolar binding affinity for F-actin, which ensures stable and reliable actin staining
  • Extensive fluorescent conjugate options—over twenty conjugated varieties of phalloidin
  • Compatibility with fixed samples—typically used with fixed cells and tissues, making them suitable for actin staining in detailed structural studies, immunofluorescence staining, and IHC applications
  • Multiplexing capability—the wide availability of phalloidin conjugates enables their use in combination with other fluorescent probes and antibodies for multiplex imaging. Biotinylated phalloidin can be made use of in downstream streptavidin steps.
  • Quantitative analysis—can be used for quantitative analysis of F-actin distribution and density within cells, aiding in the study of cytoskeletal dynamics. The unlabeled phalloidin can be titrated as a control.
  • Ease of use—staining is straightforward and quick
  • Excellent stability—exhibit good photostability, which is essential for prolonged imaging sessions and time-lapse studies
  • Wide applicability—used for a range of applications, including studying cell morphology, motility, and the effects of drugs on the actin cytoskeleton

Order Info

Shipping Condition: Room Temperature

Specifications

Color Red-orange
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Excitation Wavelength Range 540/565 nm
Dye Type TRITC
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer, Confocal Microscope, Compatible with TRITC/RFP filter set
Quantity 300 Units
Shipping Condition Room Temperature
Label Type Classic Dyes
Product Type Phalloidin
SubCellular Localization Actin, Cytoskeleton
Can I combine Click-iT or Click-iT Plus reactions with phalloidin conjugates used for actin staining?

We do not recommend using phalloidin conjugates for staining actin in combination with traditional Click-iT or Click-iT Plus reactions since phalloidin is extremely sensitive to the presence of copper.

For staining actin in combination with traditional Click-iT or Click-iT Plus reactions, we recommend using anti-α-actin antibodies for staining actin in the cytoskeleton. You can find a list of our actin antibodies here.

Another option would be to use the Click-iT Plus Alexa Fluor Picolyl Azide Toolkit (Cat. Nos. C10641, C10642, C10643). These Click-iT Plus toolkits provide Copper and Copper protectant separately which makes it easier to titrate the copper concentration to obtain optimal labeling with minimal copper-mediated damage. You may need to optimize the click reaction with the lowest possible concentration of copper and then perform the phalloidin staining.

I'm trying to label my paraffin sections for F-actin with a phalloidin conjugate, but I'm not seeing any signal. Why?

When cells and tissues are treated with solvents such as xylene or acetone (for example during deparaffinization of tissue sections), it affects the F-actin in a way that prevents phalloidins from binding. Phalloidin may be used with cryosections, which are not typically washed with organic solvents, or anti-actin antibodies may be used.

For Research Use Only. Not for use in diagnostic procedures.