missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PHACTR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA514597
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA514597 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA514597 Supplier Invitrogen™ Supplier No. PA514597
Only null left

Rabbit Polyclonal Antibody

PHACTR2 Polyclonal Antibody for Western Blot, IHC (P)

Specifications

Antigen PHACTR2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PHACTR2
Gene Accession No. O75167
Gene Alias C6orf56; KIAA0680; PHACTR2; Phosphatase and actin regulator 2
Gene Symbols PHACTR2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 169-198 amino acids from the N-terminal region of human PHACTR2.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9749
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less