missing translation for 'onlineSavingsMsg'
Learn More

pH - WS, MilliporeSigma™ Supelco™

Catalog No. p-7187409
Change view
Click to view available options
Quantity:
100 mL
20 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002650 20 mL
111002642 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111002650 Supplier Merck Emd Millipore Supplier No. PE136820ML
Only null left

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Featured Industries: Environmental

Specifications

CAS 1330-43-4
Synonym pH-WS
Type Proficiency Testing Material
Quantity 20 mL