Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
pH in Water- WP, Ready to use Standard, MilliporeSigma™ Supelco™
Description
Featured Industries: Environmental
Specifications
Specifications
| Packaging | Ampule |
| CAS | 1330-43-4 |
| Synonym | pH aq. soln.- WP |
| Type | Proficiency Testing Material |
| Quantity | 20 mL |