Learn More
Fisherbrand™ pH Test Strips
Conveniently packed in flip-top vials, these multipad pH strips provide excellent color separation over the entire pH scale with clear multicolor matches at every interval.
$50.90 - $56.36
Specifications
Range Narrow
View More Specs
Products 7
Catalog Number Mfr. No. pH Range Price Quantity & Availability  
Catalog Number Mfr. No. pH Range Price Quantity & Availability  
13640522
View Documents Promotion Details
Fisherbrand™
80060FISHER
0 to 6
Each for $50.90
Case of 6 Each for $305.40
Only null left
13640521
View Documents Promotion Details
Fisherbrand™
80014FISHER
0 to 14
Each for $56.36
Case of 6 Each for $338.16
Only null left
13640527
View Documents Promotion Details
Fisherbrand™
82090FISHER
2 to 9
Each for $50.90
Case of 6 Each for $305.40
Only null left
13640523
View Documents Promotion Details
Fisherbrand™
84070FISHER
4 to 7
Each for $50.90
Case of 6 Each for $305.40
Only null left
13640524
View Documents Promotion Details
Fisherbrand™
84510FISHER
4.5 to 10
Each for $50.90
Case of 6 Each for $305.40
Only null left
13640525
View Documents Promotion Details
Fisherbrand™
86010FISHER
6 to 10
Each for $50.90
Case of 6 Each for $305.40
Only null left
13640526
View Documents Promotion Details
Fisherbrand™
87014FISHER
7 to 14
Each for $50.90
Case of 6 Each for $305.40
Only null left
Description
Description
  • Non bleed, multiple pad color match
  • Dip and read instant color match
  • Flip-top vial stays closed when dropped, but is easy to open with one-hand
  • Sturdy vial keeps strips safe from the elements
  • 100 strips per vial
Specifications
Specifications
Narrow