missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ pH/Ion Meter SevenDirect SD50

The SevenDirect SD50 is an accurate and flexible benchtop meter for the determination of pH, ORP and ion concentrations. The instrument is the replacement of the SevenCompact and is delivered with an EasyPlace sensor arm.

Supplier:  Mettler Toledo™ 30671548

 View more versions of this product

Catalog No. 01911926


Earn Up to 5X the Fisher Scientific Buck$ with Purchase
View all promotions
Only null left

Description

Description

SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of pH, ORP and ion content of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.

  • Large color touchscreen provides direct measurement and easy data entry.
  • Instructions guide users through the sensor calibration process.
  • View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
  • The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
  • A two-level user management protects the most important settings.
  • The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
  • Dedicated positions for cleaning and storage support correct sensor handling.
Specifications

Chemical Identifiers

0.5 %
PC, Printer, USB Stick
Color TFT
pH
-2.000 to 20.000
-30°C to +130°C
-2,000 to 20,000 pX, 0 to 999,999 mg/L (ppm)
±0.1°C between 0°C and 100°C
0.1°C
0.002
1
IP54
-2000 to 2000 mV
0.1, 0.01, 0.001
2000 Standard Measurements
3 or 4 digits (auto-range)
-22°F to +266°F
195 x 205 x 65 mm

Specifications

0.5 %
0.002
SevenDirect SD50 Pure H2O
Color TFT
pH
-2.000 to 20.000
-30°C to +130°C
2000 Standard Measurements
Ready-to-measure kit
3 or 4 digits (auto-range)
1, 0.1
-22°F to +266°F
195 x 205 x 65 mm
850 g
0.1 mV
PC, Printer, USB Stick
1
IP54
-2000 to 2000 mV
0.1, 0.01, 0.001
InLab Pure-Pro ISM pH/Temperature Sensor
Pure Pro-ISM
-2,000 to 20,000 pX, 0 to 999,999 mg/L (ppm)
1, 0.1 mV
±0.1°C between 0°C and 100°C
0.1°C
1.9 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions