missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ pH Gradient Buffer Concentrates
GREENER_CHOICE

Catalog No. DX085346
Change view
Click to view available options
Includes:
CX-1 pH Gradient Buffer A
CX-1 pH Gradient Buffer B
Quantity:
125 mL
250 mL
500 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Quantity
DX085346 CX-1 pH Gradient Buffer A 250 mL
DX083273 CX-1 pH Gradient Buffer A 125 mL
DX083275 CX-1 pH Gradient Buffer B 125 mL
DX085348 CX-1 pH Gradient Buffer B 250 mL
DX302779 CX-1 pH Gradient Buffer A 500 mL
DX302780 CX-1 pH Gradient Buffer B 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. DX085346 Supplier Thermo Scientific™ Supplier No. 085346

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Robust reproducible gradients that are applicable to a wide range of MAbs and ready to use with existing LC columns and systems, without the need to formulate mobile phases.

Obtain fast and highly robust, reproducible HPLC gradients using Thermo Scientific™ CX-1 pH Gradient Buffers. Simple to optimize and easily automated, these buffers are applicable for HPLC separation of the majority of monoclonal antibody (mAb) biotherapeutics drugs. CX-1 pH Gradient Buffers are ready to use with existing HPLC columns without the need to formulate mobile phases. The CX-1 buffers save time in method development, facilitate method transfer to QA/QC for a wide range of mAb charge variants through a generic HPLC-based approach to mAb characterization, and do not require time-consuming mobile phase adjustments.

pH buffer solutions can be used to generate highly reproducible, linear pH gradients in cation exchange chromatography.

  • Robust, reproducible gradients
  • Predict biomolecule pH
  • Simple method optimization
  • Easily automated
  • Applicable to the majority of MAbs

Unlike traditional cation exchange chromatography using salt gradients, it is possible to predict the pH and the expected retention of the charge variants and use a narrow pH range to get a higher resolution separation.

Specifications

Description CX-1 pH Gradient Buffer A
For Use With (Equipment) LC Columns and Systems
Volume (Metric) 250 mL
Includes CX-1 pH Gradient Buffer A
pH 5.6
Quantity 250 mL
Standard Concentration 10X
Green Features ACT Ecolabel (Gold)
Product Line Thermo Scientific