missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ pH/Conductivity Meter SevenDirect SD23

Catalog No. 01911930
Change view
Click to view available options
Probe Type:
Not included
Pure Pro-ISM
Routine Pro-ISM
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Probe Type
01911930 Pure Pro-ISM
01911912 Not included
01911929 Routine Pro-ISM
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01911930 Supplier Mettler Toledo™ Supplier No. 30671568

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The SevenDirect SD23 is an accurate and flexible benchtop meter for the determination of pH and conductivity - either separately or simultaneously. The instrument is the replacement of the SevenCompact and is delivered with an EasyPlace sensor arm.

SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of pH, ORP and conductivity of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.

  • Large color touchscreen provides direct measurement and easy data entry.
  • Instructions guide users through the sensor calibration process.
  • View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
  • The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
  • A two-level user management protects the most important settings.
  • The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
  • Dedicated positions for cleaning and storage support correct sensor handling.

Specifications

Accuracy (Conductivity) 0.5 %
Accuracy (mV) 0.1 mV
Accuracy (pH) 0.002
Data Management Details PC, Printer, USB Stick
Description SevenDirect SD23 Pure H2O
Channels 2
Display Type Color TFT
IP Rating IP54
Range (Conductivity) 0.001 μS/cm to 2000 mS/cm
Range (mV) -2000 to 2000 mV
Resolution (pH) 0.1, 0.01, 0.001
Temperature Range (Metric) -30°C to +130°C
Includes InLab Pure-Pro ISM pH/Temperature Sensor
Memory 2000 Standard Measurements
Probe Type Pure Pro-ISM
Product Type Ready-to-measure kit
Resolution (Conductivity) 0.001 to 1
Resolution (mV) 1, 0.1 mV
Resolution (ORP) 1, 0.1
Temperature Accuracy ±0.1°C between 0°C and 100°C
Temperature Range (English) -22°F to +266°F
Temperature Resolution 0.1°C
Dimensions (L x W x H) 195 x 205 x 65 mm
Weight (English) 1.9 lb.
Weight (Metric) 850 g
Show More Show Less