missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ pH/Conductivity Meter SevenDirect SD23
GREENER_CHOICE

Catalog No. 01911912
Change view
Click to view available options
Probe Type:
Not included
Pure Pro-ISM
Routine Pro-ISM
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Probe Type
01911912 Not included
01911929 Routine Pro-ISM
01911930 Pure Pro-ISM
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01911912 Supplier Mettler Toledo™ Supplier No. 30671553
  • ONLINE EXCLUSIVE PRICE
    $3,836.07
    $2,621.31 / Each

    Save $1,214.76

    32.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The SevenDirect SD23 is an accurate and flexible benchtop meter for the determination of pH and conductivity - either separately or simultaneously. The instrument is the replacement of the SevenCompact and is delivered with an EasyPlace sensor arm.

SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of pH, ORP and conductivity of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.

  • Large color touchscreen provides direct measurement and easy data entry.
  • Instructions guide users through the sensor calibration process.
  • View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
  • The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
  • A two-level user management protects the most important settings.
  • The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
  • Dedicated positions for cleaning and storage support correct sensor handling.

Specifications

Accuracy (Conductivity) 0.5 %
Accuracy (mV) 0.1 mV
Accuracy (pH) 0.002
Data Management Details PC, Printer, USB Stick
Description SevenDirect SD23, Meter Only
Channels 2
Display Type Color TFT
IP Rating IP54
Range (Conductivity) 0.001 μS/cm to 2000 mS/cm
Range (mV) -2000 to 2000 mV
Resolution (pH) 0.1, 0.01, 0.001
Temperature Range (Metric) -30°C to +130°C
Memory 2000 Standard Measurements
Probe Type Not included
Product Type Meter only
Resolution (Conductivity) 0.001 to 1.000
Resolution (mV) 1, 0.1 mV
Resolution (ORP) 1, 0.1
Temperature Accuracy ±0.1°C between 0°C and 100°C
Temperature Range (English) -22°F to +266°F
Temperature Resolution 0.1°C
Dimensions (L x W x H) 195 x 205 x 65 mm
Weight (English) 1.9 lb.
Weight (Metric) 850 g
Show More Show Less