missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH/Conductivity - Clay Soil, MilliporeSigma™ Supelco™
Description
- pH: 6.33 ± 0.157 units
- Specific conductance, Conductivity: 900 ± 36.6 μmhos/cm
Specifications
Specifications
| CAS | 1330-43-4 |
| Type | Certified Reference Material |
| Quantity | 100 g |
| Grade | Certified Reference Material |