missing translation for 'onlineSavingsMsg'
Learn More

pH Buffer Sachets, METTLER TOLEDO™
GREENER_CHOICE

Catalog No. p-194215
Change view
Click to view available options
Quantity:
30 x 20 mL
pH:
10.0
4, 7, 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity pH
01911289 30 x 20 mL 10.0
01911291 30 x 20 mL 4, 7, 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01911289 Supplier METTLER TOLEDO Supplier No. 51302079
Only null left

Buffer solutions in sealed sachets for easy and precise calibration of pH measurement equipment.

  • Traceable to NIST primary standards

Specifications

Color Blue/White
Concentration 0.02 M
For Use With (Application) Calibration
For Use With (Equipment) pH Meter
pH 10.0
Physical Form Liquid
Quantity 30 x 20 mL
Solution Type Buffer
Packaging Satchets
Grade Laboratory