missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH Buffer Sachets, METTLER TOLEDO™

Description
- Traceable to NIST primary standards
Spécifications
Spécifications
| Color | Blue/White |
| Concentration | 0.02 M |
| For Use With (Application) | Calibration |
| For Use With (Equipment) | pH Meter |
| pH | 10.0 |
| Physical Form | Liquid |
| Quantity | 30 x 20 mL |
| Solution Type | Buffer |
| Packaging | Satchets |
| Grade | Laboratory |