missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ pH Buffer, Certified Reference Material

Catalog No. RV1505001A
Change view
Click to view available options
Quantity:
500 mL
1 L
Color:
Blue
Colorless
Red
Yellow
pH:
1.00 ±0.01
1.68 ±0.01
10.00 ±0.01
11.00 ±0.01
12.00 ±0.01
12.45 ±0.01
13.00 ±0.01
2.00 ±0.01
3.00 ±0.01
4.00 ±0.01
5.00 ±0.01
6.00 ±0.01
6.86 ±0.01
7.00 ±0.01
8.00 ±0.01
9.00 ±0.01
31 product options available for selection
Product selection table with 31 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color pH
RV1505001A 1 L Colorless 5.00 ±0.01
01185028 500 mL Colorless 1.00 ±0.01
01185027 500 mL Colorless 1.68 ±0.01
01100270 500 mL Colorless 2.00 ±0.01
01185026 500 mL Colorless 3.00 ±0.01
01185025 500 mL Colorless 4.00 ±0.01
01100272 500 mL Red 4.00 ±0.01
01185024 500 mL Colorless 5.00 ±0.01
01185023 500 mL Colorless 6.00 ±0.01
01185022 500 mL Colorless 6.86 ±0.01
01185021 500 mL Colorless 7.00 ±0.01
01100274 500 mL Yellow 7.00 ±0.01
01185020 500 mL Colorless 8.00 ±0.01
01185019 500 mL Colorless 9.00 ±0.01
01185018 500 mL Colorless 10.00 ±0.01
01100276 500 mL Blue 10.00 ±0.01
01185017 500 mL Colorless 11.00 ±0.01
01185016 500 mL Colorless 12.00 ±0.01
01100277 500 mL Colorless 12.45 ±0.01
01185015 500 mL Colorless 13.00 ±0.01
RV1495001A 1 L Colorless 3.00 ±0.01
RV1500001A 1 L Colorless 4.00 ±0.01
01100271 1 L Red 4.00 ±0.01
RV1510001A 1 L Colorless 6.00 ±0.01
RV1540001A 1 L Colorless 6.86 ±0.01
RV1550001A 1 L Colorless 7.00 ±0.01
01100273 1 L Yellow 7.00 ±0.01
RV1580001A 1 L Colorless 8.00 ±0.01
RV1590001A 1 L Colorless 9.00 ±0.01
RV1600001A 1 L Colorless 10.00 ±0.01
01100275 1 L Blue 10.00 ±0.01
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 31 options available.
31 options
Catalog No. RV1505001A Supplier VeriSpec™ Supplier No. RV1505001A

missing translation for 'validMainframeMsg'

ISO 17025/17034 pH Buffer Certified Reference Materials provide the highest level of quality assurance. A certificate of analysis providing the specified property value, uncertainty, and traceability is available with every bottle.

  • No formaldehyde or mercury compounds as preservatives
  • No chromium compounds as colorants
  • Manufactured and tested in ISO 17025/17034 accredited facilities
  • pH 4, 7, and 10 are color coded for easy identification
  • Certified traceable to NIST

Specifications

CAS 7732-18-5, 877-24-7, 1310-73-2
Color Colorless
Certificate of Analysis Yes
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 5.00 ±0.01
Physical Form Liquid
Product Line VeriSpec
Quantity 1 L
Solution Type pH Buffer
Packaging 1 L Natural Poly
Certifications/Compliance ISO 17025/17034
Grade Certified
Show More Show Less

ISO 17025/ISO 17034