missing translation for 'onlineSavingsMsg'
Learn More

Ricca VeriSpec™ pH Buffer, Certified Reference Material

Catalog No. 01100277
Change view
Click to view available options
Quantity:
500 mL
1 L
Color:
Blue
Colorless
Red
Yellow
pH:
1.00 ±0.01
1.68 ±0.01
10.00 ±0.01
11.00 ±0.01
12.00 ±0.01
12.45 ±0.01
13.00 ±0.01
2.00 ±0.01
3.00 ±0.01
4.00 ±0.01
5.00 ±0.01
6.00 ±0.01
6.86 ±0.01
7.00 ±0.01
8.00 ±0.01
9.00 ±0.01
31 product options available for selection
Product selection table with 31 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color pH
01100277 500 mL Colorless 12.45 ±0.01
01185028 500 mL Colorless 1.00 ±0.01
01185027 500 mL Colorless 1.68 ±0.01
01100270 500 mL Colorless 2.00 ±0.01
01185026 500 mL Colorless 3.00 ±0.01
01185025 500 mL Colorless 4.00 ±0.01
01100272 500 mL Red 4.00 ±0.01
01185024 500 mL Colorless 5.00 ±0.01
01185023 500 mL Colorless 6.00 ±0.01
01185022 500 mL Colorless 6.86 ±0.01
01185021 500 mL Colorless 7.00 ±0.01
01100274 500 mL Yellow 7.00 ±0.01
01185020 500 mL Colorless 8.00 ±0.01
01185019 500 mL Colorless 9.00 ±0.01
01185018 500 mL Colorless 10.00 ±0.01
01100276 500 mL Blue 10.00 ±0.01
01185017 500 mL Colorless 11.00 ±0.01
01185016 500 mL Colorless 12.00 ±0.01
01185015 500 mL Colorless 13.00 ±0.01
RV1495001A 1 L Colorless 3.00 ±0.01
RV1500001A 1 L Colorless 4.00 ±0.01
01100271 1 L Red 4.00 ±0.01
RV1505001A 1 L Colorless 5.00 ±0.01
RV1510001A 1 L Colorless 6.00 ±0.01
RV1540001A 1 L Colorless 6.86 ±0.01
RV1550001A 1 L Colorless 7.00 ±0.01
01100273 1 L Yellow 7.00 ±0.01
RV1580001A 1 L Colorless 8.00 ±0.01
RV1590001A 1 L Colorless 9.00 ±0.01
RV1600001A 1 L Colorless 10.00 ±0.01
01100275 1 L Blue 10.00 ±0.01
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 31 options available.
31 options
Catalog No. 01100277 Supplier Ricca VeriSpec™ Supplier No. RV161800500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ISO 17025/17034 pH Buffer Certified Reference Materials provide the highest level of quality assurance. A certificate of analysis providing the specified property value, uncertainty, and traceability is available with every bottle.

  • No formaldehyde or mercury compounds as preservatives
  • No chromium compounds as colorants
  • Manufactured and tested in ISO 17025/17034 accredited facilities
  • pH 4, 7, and 10 are color coded for easy identification
  • Certified traceable to NIST

Specifications

Chemical Name or Material VeriSpec™ Certified Reference Material, Buffer pH 12.45 ±0.01 (at 25°C)
Color Colorless
Certificate of Analysis Yes
CAS 7447-40-7, 1310-73-2, 7731-18-5
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 12.45 ±0.01
Physical Form Liquid
Product Line VeriSpec
Quantity 500 mL
Solution Type pH Buffer
Formulation Aqueous Solution
Packaging PETG Clear
Traceability to NIST NIST Traceable
Grade Certified
Show More Show Less

ISO 17025/ISO 17034