missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 4 Buffer Standard, SPEX CertiPrep™
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| pH | 4 SI Units |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | pH 4 Buffer Standard |
| Concentration | 4 mol/L |
| For Use With (Application) | pH Calibration |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Solution Type | Standard |
| Packaging | Triple-leached LDPE bottle |
| Concentration or Composition (by Analyte or Components) | Various |
Product intended for laboratory use only