missing translation for 'onlineSavingsMsg'
Learn More

PGRMC1 Mouse anti-Human, Clone: 108-B6, MilliporeSigma™

Mouse Monoclonal Antibody

$284.60 - $731.13

Specifications

Antigen PGRMC1
Clone 108-B6
Concentration 1.0 mg/mL
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MABF265425
View Documents
MilliporeSigma
MABF265425UG
25 μg
Each for $284.60
Only null left
 
MABF2654100
View Documents
MilliporeSigma
MABF2654100UG
100 μg
Each for $731.13
Only null left
 
Description

Description

PGRMC1 Monoclonal antibody specifically detects PGRMC1 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blotting.
Specifications

Specifications

PGRMC1
1.0 mg/mL
Monoclonal
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG1 κ
Protein G purified
108-B6
Flow Cytometry, Immunoprecipitation, Western Blot
Unconjugated
Mouse
Human
H9 cells cultured with retinoic acid.
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'