missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PGLS Polyclonal Antibody
GREENER_CHOICE

Catalog No. pipa559277
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559277 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559277 Supplier Invitrogen™ Supplier No. PA559277
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: YRTHLLSRLP IPESQVITIN PELPVEEAAE DYAKKLRQAF QGDSIPVFDL LILGVGPDGH TCSLFPDHPL LQEREQ Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.

PGLS belongs to the glucosamine/galactosamine-6-phosphate isomerase family, 6-phosphogluconolactonase subfamily. It is implicated in the hydrolysis of 6-phosphogluconolactone to 6-phosphogluconate.

Specifications

Antigen PGLS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PGLS
Gene Accession No. O95336
Gene Alias 1110030K05Rik; 6PGL; 6PGL {ECO:0000250; 6-phosphogluconolactonase; 6-phosphogluconolactonase {ECO:0000250; AI447866; epididymis secretory protein Li 304; HEL-S-304; hypothetical protein LOC616120; Pgls; pgls {ECO:0000250; Plgs; UniProtKB:O95336}
Gene Symbols PGLS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PGLS. Recombinant protein control fragment (Product #RP-99069).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25796
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less