missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega pGEM™ Vectors
Description
Comprend
Vector, 20μg; Bacterial Strain JM109, 500μL
- Standard Cloning Vectors That Can Produce ssDNA and In vitro Transcribe Sense and Antisense RNA
- Blue/white screening for easy identification of recombinant clones
- In vitro transcription using the SP6 and T7 RNA polymerase promoters flanking the multiple cloning region
- The difference between + and - vectors is orientation of the f1 origin
Spécifications
Spécifications
| Promoter | SP6, T7 |
| Restriction Site | EcoRI, SacI, KpnI, AvaI, SmaI, BamHI, XbaI, SalI, AccI, HincII, PstI, SphI and HindIII |
| Content And Storage | -30°C to -10°C |
| Quantity | 20 μg |
| Vector | pGEM-3Zf(+) vector |
| For Use With (Application) | Synthesis of RNA in-vitro and allows selection of recombinants by blue/white screening |