missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGCP Polyclonal Antibody

Description
PGCP Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | PGCP |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.82 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Cpq |
| Gene Accession No. | Q9WVJ3 |
| Gene Alias | 1190003P12Rik; 2610034C17Rik; aminopeptidase; blood plasma glutamate carboxypeptidase; carboxypeptidase Q; Cpq; Hematopoietic lineage switch 2; hematopoietic lineage switch 2 related protein; Hls2; hls2-rp; LAL; Lal-1; LCH1; LDP; liver annexin like-1; liver annexin-like protein 1; Lysosomal dipeptidase; Pgcp; Plasma glutamate carboxypeptidase; Ser-Met dipeptidase |
| Gene Symbols | Cpq |
| Show More |