Learn More
Promega pFN2K (GST) Flexi™ Vector
Catalog No. PRC8471
Encompass
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
PRC8471 20 μg
1 options
Catalog No. PRC8471 Supplier Promega Supplier No. C8471
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
These vectors are used for inducible expression of N-terminal GST-tagged fusion proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter.
  • Protein Expression Vectors with an N-terminal GST Fusion Tag
  • Express in E. coli or a cell-free expression system using the T7 promoter
  • Use N-terminal glutathione-S-transferase tag to purify or bind fusion proteins
  • Cleave GST from protein using TEV protease
Specifications
Content And Storage -30°C to -10°C
Quantity 20 μg