Learn More
Promega pF5K CMV-neo Flexi™ Vector
Catalog No. PRC9411
Encompass
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
PRC9411 20 μg
1 options
Catalog No. PRC9411 Supplier Promega Supplier No. C9411
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Flexi Vector System is a simple, yet powerful, directional cloning method for protein-coding sequences.
  • Untagged Proteins Expressed in Mammalian Cells with a Selectable Marker
  • Expresses high-levels of protein from cloned insert with CMV promoter
  • Transfer your cloned insert into other Flexi(R) Vectors for additional applications (e.g., bacterial or cell-free protein expression) as needed
  • Adapts to high-throughput formats for large screening projects, enhancing productivity
Specifications
Content And Storage -30°C to -10°C
Quantity 20 μg