missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PF4V1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150247
Description
PF4V1 Polyclonal Antibody for Western Blot, ICC/IF, IHC
The PF4V1 (Platelet Factor 4 Variant 1) gene, located on chromosome 4q12, encodes a protein that is a member of the CXC chemokine family, sharing significant homology with platelet factor 4 (PF4/CXCL4). PF4V1 is primarily expressed in megakaryocytes and platelets, playing a role in modulating coagulation and inflammatory responses. The protein has chemotactic activity, attracting immune cells to sites of injury or inflammation. Furthermore, PF4V1 can bind to heparin, influencing the anticoagulant activity of heparin and potentially impacting hemostasis and thrombosis. Research suggests that variations in the expression or function of PF4V1 might contribute to different pathological conditions, including cardiovascular disease and inflammatory disorders.Chemical Identifiers
| PF4V1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P10720 | |
| 5197 | |
| Recombinant Platelet Factor 4 Variant 1 (PF4V1) (Met1-Ser104). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| PF4V1 | |
| C-X-C motif chemokine 4; C-X-C motif chemokine 4 variant; CXCL4L1; CXCL4V1; PF4A; PF4alt; PF4-ALT; PF4V1; PF4var1; platelet factor 4 variant; platelet factor 4 variant 1; Platelet factor 4 variant(4-74); Platelet factor 4 variant(5-74); Platelet factor 4 variant(6-74); platelet factor 4, variant 1 (PF4-like); SCYB4V1 | |
| PF4V1 | |
| Primary | |
| Protein A |
Specifications
| PF4V1 | |
| Polyclonal | |
| Unconjugated | |
| PF4V1 | |
| C-X-C motif chemokine 4; C-X-C motif chemokine 4 variant; CXCL4L1; CXCL4V1; PF4A; PF4alt; PF4-ALT; PF4V1; PF4var1; platelet factor 4 variant; platelet factor 4 variant 1; Platelet factor 4 variant(4-74); Platelet factor 4 variant(5-74); Platelet factor 4 variant(6-74); platelet factor 4, variant 1 (PF4-like); SCYB4V1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5197 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P10720 | |
| PF4V1 | |
| Recombinant Platelet Factor 4 Variant 1 (PF4V1) (Met1-Ser104). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |