Learn More
Promega pF4A CMV Flexi™ Vector
Catalog No. PRC8481
Encompass
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
PRC8481 20 μg
1 options
Catalog No. PRC8481 Supplier Promega Supplier No. C8481
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Designed for high-level expression of proteins in mammalian cells from the CMV promoter without a selectable marker
  • Untagged Proteins Expressed in Mammalian Cells Without a Selectable Marker
  • Expresses high-levels of protein from cloned insert with CMV promoter
  • Transfer your cloned insert into other Flexi(R) Vectors for additional applications (e.g., bacterial or cell-free protein expression) as needed
  • Adapts to high-throughput formats for large screening projects, enhancing productivity
Specifications
Content And Storage -30°C to -10°C
Quantity 20 μg