Learn More
Promega pF1K T7 Flexi™ Vector
Catalog No. PRC8451
Encompass
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
PRC8451 20 μg
1 options
Catalog No. PRC8451 Supplier Promega Supplier No. C8451
These untagged vectors are used for inducible expression of proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter. Supplied as vectors used with the Flexi Cloning System.
  • Untagged Bacterial Expression Vectors
  • Generate protein from T7 promoter in E. coli and cell-free expression systems
  • Options for Kanamycin and Ampicillin resistance
  • Starting vector for transferring ORFs into other Flexi(R) Vectors
Specifications
Content And Storage -30°C to -10°C
Quantity 20 μg