missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150017
Description
PEX1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a member of the AAA ATPase family, a large group of ATPases associated with diverse cellular activities. This protein is cytoplasmic but is often anchored to a peroxisomal membrane where it forms a heteromeric complex and plays a role in the import of proteins into peroxisomes and peroxisome biogenesis. Mutations in this gene have been associated with complementation group 1 peroxisomal disorders such as neonatal adrenoleukodystrophy, infantile Refsum disease, and Zellweger syndrome. Alternatively spliced transcript variants have been found for this gene.Chemical Identifiers
| PEX1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q5BL07 | |
| 500006, 71382 | |
| Recombinant Peroxisomal Biogenesis Factor 1 (PEX1) (Asn172-Ala377). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| Pex1 | |
| 5430414H02Rik; E330005K07Rik; HMLR1; PBD1A; PBD1B; Peroxin-1; peroxisomal biogenesis factor 1; peroxisome biogenesis disorder protein 1; Peroxisome biogenesis factor 1; Pex1; RGD1559939; Zellweger syndrome; ZWS; ZWS1 | |
| Pex1 | |
| Primary | |
| Protein A |
Specifications
| PEX1 | |
| Polyclonal | |
| Unconjugated | |
| Pex1 | |
| 5430414H02Rik; E330005K07Rik; HMLR1; PBD1A; PBD1B; Peroxin-1; peroxisomal biogenesis factor 1; peroxisome biogenesis disorder protein 1; Peroxisome biogenesis factor 1; Pex1; RGD1559939; Zellweger syndrome; ZWS; ZWS1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 500006, 71382 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q5BL07 | |
| Pex1 | |
| Recombinant Peroxisomal Biogenesis Factor 1 (PEX1) (Asn172-Ala377). | |
| 200 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody |