Learn More
Justrite PetroPad
Reusable, washable and easy to dispose absorbent pad designed for the safe handling and the disposal of hazardous hydrocarbon waste.
Supplier:  Justrite 83984
 View more versions of this product
Catalog No. 17988515
Only null left
Description
Description
  • Portable and easy to fold up and store without fear of leakage or mess
  • Absorbs and permanently solidifies hydrocarbons while allowing rainwater to pass through
  • Durable material and robust construction withstand the demands of a rigorous work environment
  • Easily pressure wash built-up dirt and debris without releasing hydrocarbons
  • TCLP leachate test and EPA Paint Filter Test (9095B) compliant
Specifications
Chemical Identifiers
Polymer
Sorbent, Oil-only
Reusable
Heavyweight
Specifications
Polymer
Reusable
35.4 x 27.2 in.
Sorbent, Oil-only
Heavyweight
Product Suggestions
Product Suggestions