Learn More
Justrite PetroPad
Reusable, washable and easy to dispose absorbent pad designed for the safe handling and the disposal of hazardous hydrocarbon waste.
Supplier: Justrite 83990
Description
- Portable and easy to fold up and store without fear of leakage or mess
- Absorbs and permanently solidifies hydrocarbons while allowing rainwater to pass through
- Durable material and robust construction withstand the demands of a rigorous work environment
- Easily pressure wash built-up dirt and debris without releasing hydrocarbons
- TCLP leachate test and EPA Paint Filter Test (9095B) compliant
Chemical Identifiers
| Polymer | |
| Sorbent, Oil-only | |
| Reusable | |
| Heavyweight |
Specifications
| Polymer | |
| Reusable | |
| 85.8 x 53.9 in. |
| Sorbent, Oil-only | |
| Heavyweight |