missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, puriss. p.a., Reag. Ph. Eur., Honeywell™

Catalog No. p-7073764
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60048547 1 L
60048548 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60048547 Supplier Honeywell Inc Supplier No. 322471L
Only null left

Chemical Identifiers

CAS 101316-46-5
MDL Number MFCD00081849

Specifications

Density 0.661 to 0.664g/mL at 20°C
Quantity 1 L
Flash Point −26°C (−14.8°F)
UN Number 1268
Grade puriss. p.a.
Chemical Name or Material Petroleum Ether, Reag. Ph. Eur.
Hazard Category Danger
Hazard Statement P201-P210-P301 + P310-P331-P370 + P378-P501
Precautionary Statement H225-H304-H315-H336-H361f-H373-H411