missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether, puriss. p.a., ACS Reagent, Reag. ISO, Solstice
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
| Appearance | Liquid |
| Density | 0.640 to 0.655g/mL at 20°C |
| Quantity | 5 L |
| Flash Point | −30°C (−22°F) |
| Packaging | Aluminum Bottle |
| Residue after Evaporation | 0 to 0.001% |
| Titratable Acid | Free acid (as CH3COOH): ≤0.002% |
| UN Number | UN1268 |
| Boiling Point | 40°C to 60°C |
| Grade | ACS/puriss. p.a. |
| Show More |
Safety and Handling
| Hazard Statement | P210-P261-P273-P301 + P310-P331 |
| Precautionary Statement | H225-H302-H304-H315-H336-H411 |
| DOTInformation | Class 3 - PG 2 - EHS - VP > 110kPa - Petroleum distillates, n.o.s., HI: S |