missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether, Purified (Ligroin), Reagents
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 35°C to 60°C |
| Quantity | 4 L |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Specific Gravity | 0.645 |
| Grade | Pure |
| Chemical Name or Material | Petroleum Ether, Purified (Ligroin) |