missing translation for 'onlineSavingsMsg'
Learn More

Petroleum ether, For pesticide residue analysis, low boiling point hydrogen treated naphtha, bp 40-60 °C, Honeywell Riedel-de Haën™

Catalog No. p-7072104
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001177 1 L
6001178 2.5 L
6001179 6 x 1 L
60046350 4 x 2.5 L
6001180 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6001177 Supplier Honeywell Chemicals Supplier No. 344911L
Only null left

CHROMASOLV™, for pesticide residue analysis, low boiling point hydrogen treated naphtha, bp 40-60 °C

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Chemical Identifiers

CAS 101316-46-5 , 109-66-0
MDL Number MFCD00081849

Specifications

Quantity 1 L
Flash Point −40°C (−40°F)
UN Number UN1268
Grade CHROMASOLV
Chemical Name or Material Petroleum ether
Hazard Statement P210-P260-P280-P301 + P310-P370 + P378-P403 + P235