missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum ether, For pesticide residue analysis, low boiling point hydrogen treated naphtha, bp 40-60 °C, Honeywell Riedel-de Haën™
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Specifications
Specifications
| CAS Min % | ≥25.0000% |
| CAS Max % | <50.0000% |
| Density | 0.65 g/cm3 |
| Quantity | 4 x 4 L |
| Flash Point | −30°C |
| Packaging | Glass Bottle |
| UN Number | UN3295 |
| Boiling Point | 40°C to 60°C |
| Grade | Pesticide |
| Chemical Name or Material | Petroleum ether, For pesticide residue analysis, low boiling point hydrogen treated naphtha, bp 40-60 °C |
Safety and Handling
| ShelfLife | 1620 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Hydrocarbons, liquid, n.o.s. |
| EINECSNumber | 309-852-0 |
| Recommended Storage | Room Temp |