missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum ether, For pesticide residue analysis, low boiling point hydrogen treated naphtha, bp 40-60 °C, Solstice
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Specifications
Specifications
| Quantity | 6 x 1 L |
| Flash Point | −40°C (−40°F) |
| UN Number | UN1268 |
| Grade | CHROMASOLV |
| Chemical Name or Material | Petroleum ether |
Safety and Handling
| Hazard Statement | P210-P260-P280-P301 + P310-P370 + P378-P403 + P235 |