missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether (Certified ACS), Fisher Chemical™

Catalog No. E139500
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Aluminum Can
Glass Bottle
Steel Drum
Steel Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
E139500 500 mL Glass Bottle
E1391 1 L Glass Bottle
E139S4 4 L Aluminum Can
E1394 4 L Glass Bottle
E13920 20 L Steel Pail
E139200 200 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. E139500 Supplier Thermo Fisher Scientific Supplier No. E139 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Also meets specifications of the American Oil Chemists Society and the National Cottonseed Producers Association

Chemical Identifiers

CAS 8032-32-4
MDL Number MFCD00081849
Synonym Ligroine, Benzine, Naphtha Petroleum, Naphtha Solvent (Optima/Pesticide/Certified ACS), magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide

Specifications

Color Colorless
Acidity Pass Test
Quantity 500 mL
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.001% max.
Specific Gravity 0.630 to 0.660 (at 25°C)
Boiling Point 38°C
Grade ACS
Chemical Name or Material Petroleum Ether
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause heritable genetic damage. May cause eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid