missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, B.R. 60 DEG -90°C, Spectrum™ Chemical

Catalog No. 18612205
Change view
Click to view available options
Quantity:
4 L
200 L
Packaging:
Amber Glass Bottle
Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612205 4 L Amber Glass Bottle
18612207 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18612205 Supplier Spectrum Chemical Mfg Cor Supplier No. P10504LTGL

Chemical Identifiers

CAS 8032-32-4

Specifications

Acidity To Pass Test
CAS Max % 1
Quantity 4 L
Odor To Pass Test
Packaging Amber Glass Bottle
Residue after Evaporation 1e-05
Boiling Point 20°C to 40°C
Grade Reagent
Chemical Name or Material Petroleum Ether

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.