missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether, B.R. 35 DEG -60°C, ACS, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor P10454LTGL
Specifications
| Petroleum Ether | |
| 1 | |
| 20°C to 40°C | |
| Amber Glass Bottle | |
| Reagent |
| 8032-32-4 | |
| To Pass Test | |
| 4 L | |
| 1e-05 |
Chemical Identifiers
| 8032-32-4 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.