missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0% (C5 & C6 Isomers), Honeywell

Catalog No. p-7112050
Change view
Click to view available options
Quantity:
19 L
4 x 4 L
Packaging:
Glass Bottle
Metal Can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001176 4 x 4 L Glass Bottle
6001175 19 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001176 Supplier Honeywell Chemicals Supplier No. LP3174
Only null left

Chemical Identifiers

CAS 8032-32-4
Synonym p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide

Specifications

CAS Max % 100.0000%
Density 0.64 g/cm3
Quantity 4 x 4 L
Flash Point −18°C
Packaging Glass Bottle
UN Number UN1268
Water 0.02% max.
Boiling Point 35°C to 60°C
Percent Purity >99%
Grade ACS
Chemical Name or Material Petroleum Ether, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0% (C5 & C6 Isomers)
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Petroleum distillates, n.o.s.