missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0% (C5 and C6 Isomers), Solstice
Specifications
Specifications
| CAS Max % | 100.0000% |
| Density | 0.64 g/cm3 |
| Quantity | 19 L |
| Flash Point | −18°C |
| Packaging | Metal Can |
| Water | 0.02% max. |
| UN Number | UN1268 |
| Boiling Point | 35°C to 60°C |
| Percent Purity | >99% |
| Grade | ACS |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Petroleum distillates, n.o.s. |