missing translation for 'onlineSavingsMsg'
Learn More

Petroleum ether 35/60, HPLC Grade

Catalog No. AA38985K2
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38985K2 1 L
38985M 4 x 1 L
AA38985K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA38985K2 Supplier Thermo Scientific Chemicals Supplier No. 038985K2

CAS: 8032-32-4

Petroleum ether 35/60 is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 8032-32-4
MDL Number MFCD00081849
Synonym p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide

Specifications

Density 0.640 g/mL
Quantity 1 L
Flash Point -49°C (-57°F)
Physical Form Liquid
Refractive Index 1.363
Boiling Point 35°C to 60°C
Solubility Information Insoluble in water.
Grade HPLC
Chemical Name or Material Petroleum ether 35/60, HPLC Grade
Hazard Category H225-H304-H315-H319-H336
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 232-453-7
RTECSNumber OI6180000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only