Learn More
Petroleum ether 35/60, ACS
Maximum level of impurities: Color (APHA): 10, Boiling range 35-60°C, Evaporation residue: 0.001%, Acidity P.T. | CAS: 8032-32-4
Supplier: Thermo Scientific Chemicals 042085M4
Description
Petroleum ether is mostly used by pharmaceutical companies and in the manufacturing process. It is a lightweight hydrocarbon used chiefly as a nonpolar solvent. The extract of aerial parts of ageratum conyzoids in petroleum ether plays an essential role for the isolation of sigmasterol and beta sitosterol. Crude petroleum spirit and methanol extracts also play an important role for the screening of antimicrobial activity of the stem bark, leaves and roots of ficus thoningii.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Petroleum ether 35/60 | |
| Colorless | |
| 0.640 g/cm3 at 20°C | |
| 20 L | |
| 250°C (482°F) | |
| Liquid | |
| MFCD00081849 | |
| p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide | |
| ACS |
| 8032-32-4 | |
| Colorless liquid | |
| 35°C to 60°C | |
| -49°C (-57°F) | |
| Petroleum-like | |
| 1.363 | |
| 350 hPa (263mm Hg) at 20°C | |
| Insoluble in water. |
Chemical Identifiers
| 8032-32-4 | |
| MFCD00081849 | |
| p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336
DOTInformation : Hazard Class: 3; Packaging Group: II
EINECSNumber : 232-453-7
RTECSNumber : OI6180000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only