missing translation for 'onlineSavingsMsg'
Learn More

Petrolatum, White, USP, Spectrum™ Chemical

Catalog No. p-7157966
Change view
Click to view available options
Quantity:
2.5 kg
15.9 kg
174 kg
Packaging:
Poly Bottle
Poly Pail
Steel Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605048 2.5 kg Poly Bottle
18605049 15.9 kg Poly Pail
18605050 174 kg Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18605048 Supplier Spectrum Chemical Mfg Cor Supplier No. PE1252.5KG
Only null left

PE125, 8009-03-8

Spectrum™ Chemical Petrolatum, White, USP is used as a moisturizer to treat or prevent dry, rough, scaly, itchy skin and minor skin irritations. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8009-03-8
Molecular Formula C15H15N
Molecular Weight (g/mol) 209.29
InChI Key WJZSZXCWMATYFX-UHFFFAOYSA-N
IUPAC Name 1,1,2-trimethyl-1H-benzo[e]indole
SMILES CC1=NC2=CC=C3C=CC=CC3=C2C1(C)C

Specifications

CAS Max % 1
Ignition Residue 0.0005
Melting Range 38-60°C
Packaging Poly Bottle
Quantity 2.5 kg
Specific Gravity 0.815-0.880
Grade USP