missing translation for 'onlineSavingsMsg'
Learn More

Petrol, puriss., 100° to 140°C, Honeywell™

Catalog No. p-7120657
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045637 1 L Glass Bottle
60045638 2.5 L Glass Bottle
60045640 4 x 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60045637 Supplier Honeywell Chemicals Supplier No. 245341L
Only null left

Puriss., bp 100-140 °C

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.

Chemical Identifiers

CAS 8032-32-4
MDL Number MFCD00081849
Synonym p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide

Specifications

Density 0.720 to 0.745g/mL at 20°C
Boiling Point 100°C to 140°C
Flash Point 10°C (50°F)
Packaging Glass Bottle
Quantity 1 L
UN Number UN3295
Grade puriss.
Chemical Name or Material Petrol
Hazard Statement P210-P261-P273-P301 + P310-P331
Precautionary Statement H225-H304-H336-H411
DOTInformation Class 3 - PG 2 - EHS - VP ≤ 110kPa - Hydrocarbons, liquid, n.o.s., HI: A, S (not BR)
EINECSNumber 920-750-0
RTECSNumber OI6180000