missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petrol, puriss., 100° to 140°C, Solstice
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Specifications
| Density | 0.720 to 0.745g/mL at 20°C |
| Boiling Point | 100°C to 140°C |
| Flash Point | 10°C (50°F) |
| Packaging | Glass Bottle |
| Quantity | 2.5 L |
| UN Number | UN3295 |
| Grade | puriss. |
| Chemical Name or Material | Petrol |
Safety and Handling
| Hazard Statement | P210-P261-P273-P301 + P310-P331 |
| Precautionary Statement | H225-H304-H336-H411 |
| DOTInformation | Class 3 - PG 2 - EHS - VP ≤ 110kPa - Hydrocarbons, liquid, n.o.s., HI: A, S (not BR) |
| EINECSNumber | 920-750-0 |
| RTECSNumber | OI6180000 |