missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Petri Dish Platinum Electrode Chamber
Petri Dish Platinum Electrode Chamber for ex vivo tissues gene or drug delivery.
$3757.90 - $3757.90
Specifications
| For Use With (Equipment) | ECM™ 830, ECM™ 2001+, Gemini X2, Legacy ECM™ 2001 |
|---|---|
| Material | Glass, Platinum |
| Product Line | BTX Electroporation |
| Product Type | Electroporation accessory |
| Includes | Petri Dish Electrode Chamber Only |
Description
- Simple and easy to use
- Available in 5 and 15 mm
- Reusable, lab-grade Pyrex glass
Specifications
| ECM™ 830, ECM™ 2001+, Gemini X2, Legacy ECM™ 2001 | |
| BTX Electroporation | |
| Petri Dish Electrode Chamber Only | |
| 5 mm | |
| Banana Plug to Micrograbber |
| Glass, Platinum | |
| Electroporation accessory | |
| 0.2 in. | |
| Non-sterile | |
| Square Wave |